Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FDFT1 SIRNA Assays ›
Gene Symbol:
FDFT1
Gene Name:
farnesyl-diphosphate farnesyltransferase 1
Gene Aliases:
DGPT, ERG9, SQS, SS
Chromosome Location:
Chr.8: 11795573 - 11839309 on Build GRCh38
UniGene ID:
Hs.593928
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
111452
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FDFT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001287742.1
NP_001274671.1
7, 8
1421
NM_001287743.1
NP_001274672.1
6, 7
1223
NM_001287744.1
NP_001274673.1
5, 6
1050
NM_001287745.1
NP_001274674.1
6, 7
1129
NM_001287747.1
NP_001274676.1
5, 6
1086
NM_001287748.1
NP_001274677.1
5, 6
748
NM_001287749.1
NP_001274678.1
5, 6
986
NM_001287750.1
NP_001274679.1
4, 5
988
NM_001287751.1
NP_001274680.1
4, 5
860
NM_001287756.1
NP_001274685.1
2, 3
445
NM_004462.4
NP_004453.3
5, 6
932
XM_017013217.1
XP_016868706.1
7, 8
1713
GenBank mRNA
AK057726.1
-
5, 6
1050
AK098682.1
-
510
AK293545.1
-
5, 6
986
AK296043.1
-
4, 5
675
AK297868.1
-
4, 5
931
AK300059.1
-
4, 5
750
AK301617.1
-
2, 3
306
AK311246.1
-
4, 5
699
AK315993.1
-
6, 7
999
AK316033.1
-
5, 6
953
AK316182.1
-
4, 5
948
AK316351.1
-
5, 6
713
AK316531.1
-
5, 6
698
AK316534.1
-
4, 5
860
BC003573.1
-
5, 6
779
3538137
BC009251.2
-
5, 6
779
2961623
BC029641.1
-
5, 6
742
4514761
BC034440.1
-
5, 6
742
BT006704.1
-
5, 6
710
CR457033.1
-
5, 6
710
L06070.1
-
5, 6
719
L06105.1
-
5, 6
754
S76822.1
-
5, 6
775
X69141.1
-
5, 6
801
Back To Top
More Information
Panther Classification:
Molecular Function -
transferase
Gene Ontology Categories:
Function(s)
Process(es)
steroid biosynthetic process
cholesterol biosynthetic process
isoprenoid biosynthetic process
farnesyl diphosphate metabolic process
oxidation-reduction process
farnesyl-diphosphate farnesyltransferase activity
oxidoreductase activity
squalene synthase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0