Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: UST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005715.2 | NP_005706.1 | 6 | 1035 | ||
GenBank mRNA | AB020316.1 | - | 6 | 842 | ||
AK292922.1 | - | 6 | 783 | |||
AK315320.1 | - | 6 | 743 | |||
BC093668.1 | - | 6 | 787 | 7939513 | ||
BC093694.1 | - | 6 | 787 | 7939539 |