Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TRIT1 SIRNA Assays ›
          Gene Symbol:
          TRIT1
          Gene Name:
          tRNA isopentenyltransferase 1
          Gene Aliases:
          GRO1, IPPT, IPT, IPTase, MOD5, hGRO1
          Chromosome Location:
          Chr.1: 39841031 - 39883511 on Build GRCh38
          UniGene ID:
          Hs.356554
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 112010

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TRIT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001312692.1 NP_001299621.1 5, 6 697
          NM_017646.5 NP_060116.2 7, 8 943
          NR_132401.1 7, 8 939
          NR_132402.1 6, 7 797
          NR_132403.1 6, 7 793
          NR_132404.1 6, 7 793
          NR_132405.1 6, 7 789
          NR_132407.1 4, 5 557
          NR_132408.1 4, 5 553
          NR_132409.1 3, 4 414
          NR_132415.1 7, 8 1030
          XM_005270954.2 XP_005271011.1 7, 8 1179
          XM_006710706.1 XP_006710769.1 7, 8 1011
          GenBank mRNA AF074918.1 - 7, 8 1024
          AK000068.1 - 5, 6 560
          AK074222.1 - 5, 6 560
          AK223478.1 - 7, 8 936
          AK307910.1 - 7, 8 928
          AY303390.1 - 7, 8 937
          AY702933.1 - 7, 8 866
          AY702934.1 - 7, 8 862
          AY702935.1 - 6, 7 713
          AY702936.1 - 6, 7 720
          AY702937.1 - 6, 7 716
          AY702938.1 - 6, 7 716
          AY702939.1 - 6, 7 712
          AY702941.1 - 4, 5 480
          AY702942.1 - 4, 5 476
          AY702943.1 - 3, 4 337
          BC010741.1 - 5, 6 659
          BC107569.1 - 7, 8 942 4594069
          BC128155.1 - 7, 8 937 40112695
          BC128156.1 - 7, 8 1024 40112698
          BE294565.1 - 4, 5 560
          CR457226.1 - 4, 5 494

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_025873, BC051040, BC019812, AK010176, AK003556, and others.

          Panther Classification:

          Molecular Function -

          RNA processing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA modification
          mitochondrial tRNA modification
          nucleic acid binding
          ATP binding
          zinc ion binding
          tRNA dimethylallyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-hdb68:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0