Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CPT1C SIRNA Assays ›
Gene Symbol:
CPT1C
Gene Name:
carnitine palmitoyltransferase 1C
Gene Aliases:
CATL1, CPT1-B, CPT1P, CPTI-B, CPTIC, SPG73
Chromosome Location:
Chr.19: 49690672 - 49713731 on Build GRCh38
UniGene ID:
Hs.112195
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
112377
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CPT1C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136052.2
NP_001129524.1
19, 20
2507
NM_001199752.2
NP_001186681.1
19, 20
2604
NM_001199753.1
NP_001186682.1
19, 20
2540
NM_152359.2
NP_689572.1
18, 19
2471
NR_108072.1
20, 21
2702
XM_005258505.2
XP_005258562.1
19, 20
2577
XM_005258506.4
XP_005258563.1
18, 19
2493
XM_006723009.3
XP_006723072.1
16, 17
2309
XM_011526438.2
XP_011524740.1
19, 20
2633
XM_011526439.2
XP_011524741.1
19, 20
2697
XM_011526440.2
XP_011524742.1
18, 19
2414
XM_017026265.1
XP_016881754.1
18, 19
2461
XM_017026266.1
XP_016881755.1
18, 19
2382
XM_017026267.1
XP_016881756.1
18, 19
2367
XM_017026268.1
XP_016881757.1
17, 18
2210
XM_017026269.1
XP_016881758.1
17, 18
2320
XM_017026270.1
XP_016881759.1
17, 18
2088
XM_017026271.1
XP_016881760.1
17, 18
2177
XM_017026272.1
XP_016881761.1
17, 18
2269
XM_017026273.1
XP_016881762.1
17, 18
2254
GenBank mRNA
AF357970.1
-
18, 19
2429
AK074389.1
-
11, 12
1450
AK289713.1
-
19, 20
2596
AK290092.1
-
19, 20
2512
AL831876.1
-
7, 8
835
BC029104.1
-
19, 20
2471
3872727
Back To Top
More Information
Panther Classification:
Molecular Function -
acyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid beta-oxidation
carnitine metabolic process
carnitine O-palmitoyltransferase activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-rlpgz:80/100.66.72.8:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0