Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TNC SIRNA Assays ›
          Gene Symbol:
          TNC
          Gene Name:
          tenascin C
          Gene Aliases:
          150-225, DFNA56, GMEM, GP, HXB, JI, TN, TN-C
          Chromosome Location:
          Chr.9: 115019575 - 115118257 on Build GRCh38
          UniGene ID:
          Hs.143250
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 11255

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TNC
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002160.3 NP_002151.2 28 7076
          XM_005251972.3 XP_005252029.1 27 6753
          XM_005251973.3 XP_005252030.1 24 5934
          XM_005251974.3 XP_005252031.1 22 5388
          XM_005251975.3 XP_005252032.1 21 5115
          XM_006717096.3 XP_006717159.1 29 7302
          XM_006717097.3 XP_006717160.1 27 6753
          XM_006717098.3 XP_006717161.1 26 6480
          XM_006717101.3 XP_006717164.1 22 5388
          XM_011518625.2 XP_011516927.1 26 6480
          XM_011518626.2 XP_011516928.1 25 6210
          XM_011518628.2 XP_011516930.1 23 5661
          XM_011518629.2 XP_011516931.1 23 5661
          XM_017014678.1 XP_016870167.1 30 7575
          XM_017014679.1 XP_016870168.1 29 7302
          XM_017014680.1 XP_016870169.1 29 7299
          XM_017014681.1 XP_016870170.1 26 6483
          GenBank mRNA AB210038.1 - 28 6952
          BC151843.1 - 25 6125 8860362
          M55618.1 - 27 6805
          X56160.1 - 27 6715
          X78565.1 - 28 6980

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          extracellular matrix protein

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          cell adhesion
          negative regulation of cell adhesion
          neuromuscular junction development
          positive regulation of cell proliferation
          response to wounding
          response to mechanical stimulus
          positive regulation of gene expression
          peripheral nervous system axon regeneration
          extracellular matrix organization
          wound healing
          odontogenesis of dentin-containing tooth
          response to ethanol
          bud outgrowth involved in lung branching
          mesenchymal-epithelial cell signaling involved in prostate gland development
          prostate gland epithelium morphogenesis
          cellular response to retinoic acid
          cellular response to vitamin D
          response to fibroblast growth factor
          cellular response to prostaglandin D stimulus
          syndecan binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0