Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HGF SIRNA Assays ›
          Gene Symbol:
          HGF
          Gene Name:
          hepatocyte growth factor
          Gene Aliases:
          DFNB39, F-TCF, HGFB, HPTA, SF
          Chromosome Location:
          Chr.7: 81699006 - 81770438 on Build GRCh38
          UniGene ID:
          Hs.396530
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 112632

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HGF
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000601.5 NP_000592.3 2 439
          NM_001010931.2 NP_001010931.1 2 439
          NM_001010932.2 NP_001010932.1 2 439
          NM_001010933.2 NP_001010933.1 2 439
          NM_001010934.2 NP_001010934.1 2 439
          XM_006715956.2 XP_006716019.1 3 402
          XM_011516115.2 XP_011514417.1 3 402
          XM_017012097.1 XP_016867586.1 3 402
          XM_017012098.1 XP_016867587.1 3 402
          GenBank mRNA AB208900.1 - 2 346
          AK291674.1 - 2 374
          AK313694.1 - 2 289
          BC022308.1 - 2 288
          BC063485.1 - 2 297
          BC105797.1 - 2 379
          BC130284.1 - 2 287 40146315
          BC130286.1 - 2 287 40146317
          BU533235.1 - 378
          BX495370.1 - 2 440
          FJ830861.1 - 2 212
          FJ830862.1 - 2 212
          L02931.1 - 2 272
          M29145.1 - 2 313
          M55379.1 - 2 212
          M60718.1 - 2 284
          M73239.1 - 2 277
          M73240.1 - 2 277
          M77227.1 - 2 294
          U46010.1 - 2 266
          X16323.1 - 2 346
          X57574.1 - 2 289

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          epithelial to mesenchymal transition
          liver development
          platelet degranulation
          proteolysis
          mitotic nuclear division
          positive regulation of cell proliferation
          negative regulation of autophagy
          positive regulation of phosphatidylinositol 3-kinase signaling
          hyaluronan metabolic process
          positive regulation of cell migration
          organ regeneration
          positive regulation of myelination
          negative regulation of interleukin-6 production
          positive regulation of interleukin-10 production
          negative regulation of peptidyl-serine phosphorylation
          cellular response to hepatocyte growth factor stimulus
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of osteoblast differentiation
          positive regulation of angiogenesis
          positive regulation of transcription from RNA polymerase II promoter
          hepatocyte growth factor receptor signaling pathway
          negative regulation of inflammatory response
          positive regulation of peptidyl-tyrosine phosphorylation
          positive chemotaxis
          myoblast proliferation
          cell chemotaxis
          regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling
          positive regulation of neuron projection regeneration
          negative regulation of release of cytochrome c from mitochondria
          regulation of p38MAPK cascade
          negative regulation of hydrogen peroxide-mediated programmed cell death
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          regulation of tau-protein kinase activity
          positive regulation of DNA biosynthetic process
          serine-type endopeptidase activity
          protein binding
          growth factor activity
          chemoattractant activity
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0