Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IL15RA SIRNA Assays ›
Gene Symbol:
IL15RA
Gene Name:
interleukin 15 receptor subunit alpha
Gene Aliases:
CD215
Chromosome Location:
Chr.10: 5948897 - 5978741 on Build GRCh38
UniGene ID:
Hs.445124
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
11274
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IL15RA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243539.1
NP_001230468.1
7
1694
NM_001256765.1
NP_001243694.1
8
1643
NM_002189.3
NP_002180.1
7
1490
NM_172200.2
NP_751950.2
6
1391
NR_046362.1
5
1400
XM_011519461.2
XP_011517763.1
9
1408
XM_011519462.2
XP_011517764.1
8
1326
XM_011519463.2
XP_011517765.1
8
1309
XM_011519464.2
XP_011517766.1
8
1255
XM_011519465.1
XP_011517767.1
9
1884
XM_011519466.2
XP_011517768.1
9
2004
XM_011519467.2
XP_011517769.1
7
1171
XM_011519469.1
XP_011517771.1
8
1785
XM_011519470.2
XP_011517772.1
6
1074
XM_011519471.2
XP_011517773.1
7
1139
XM_011519472.2
XP_011517774.1
6
978
XM_011519474.2
XP_011517776.1
6
961
XM_011519475.2
XP_011517777.1
5
879
XM_011519476.2
XP_011517778.1
5
1345
XM_011519477.2
XP_011517779.1
5
1470
XM_017016195.1
XP_016871684.1
8
1731
XM_017016196.1
XP_016871685.1
7
980
XM_017016197.1
XP_016871686.1
8
995
XM_017016198.1
XP_016871687.1
7
1615
XM_017016200.1
XP_016871689.1
6
1566
XM_017016201.1
XP_016871690.1
7
1859
XM_017016202.1
XP_016871691.1
8
1580
XM_017016203.1
XP_016871692.1
6
2042
GenBank mRNA
AF035279.1
-
7
1425
BC065506.1
-
5
1392
BC107777.1
-
7
1558
6497994
U31628.1
-
7
1453
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
cell proliferation
cytokine-mediated signaling pathway
signal transducer activity
cytokine receptor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0