Search
Search

CPB2| Gene Symbol: CPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278541.1 | NP_001265470.1 | 8 | 906 | ||
| NM_001872.4 | NP_001863.3 | 9 | 1017 | |||
| XM_017020393.1 | XP_016875882.1 | 9 | 960 | |||
| GenBank mRNA | AB011969.1 | - | 8 | 858 | ||
| AK290829.1 | - | 9 | 965 | |||
| BC007057.1 | - | 9 | 974 | 3934520 | ||
| BT006936.1 | - | 9 | 950 | |||
| M75106.1 | - | 9 | 969 | |||