Search
Search

AFG3L2| Gene Symbol: AFG3L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006796.2 | NP_006787.2 | 7 | 911 | ||
| XM_011525601.2 | XP_011523903.1 | 7 | 944 | |||
| GenBank mRNA | BC024282.1 | - | 8 | 764 | ||
| BC065016.1 | - | 7 | 833 | 6169804 | ||
| Y18314.1 | - | 7 | 830 | |||