Search
Search

CPA6| Gene Symbol: CPA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020361.4 | NP_065094.3 | 3 | 476 | ||
| XM_017013647.1 | XP_016869136.1 | 3 | 1205 | |||
| GenBank mRNA | AF221594.1 | - | 3 | 433 | ||
| BC033684.1 | - | 3 | 436 | 5193403 | ||
| BI756197.1 | - | 3 | 476 | |||