Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: C1QTNF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015645.4 | NP_056460.1 | 13 | 2223 |
Gene Symbol: MFRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031433.3 | NP_113621.1 | 13 | 2223 | ||
GenBank mRNA | AB055505.1 | - | 13 | 2210 | ||
AJ862823.1 | - | 13 | 2205 |