Search
Search

ADAMTS19| Gene Symbol: ADAMTS19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011543246.2 | XP_011541548.1 | 3 | 303 | ||
| XM_011543247.2 | XP_011541549.1 | 3 | 268 | |||
| XM_011543248.2 | XP_011541550.1 | 3 | 388 | |||
| XM_017009174.1 | XP_016864663.1 | 2 | 1062 | |||
| XM_017009176.1 | XP_016864665.1 | 3 | 268 | |||
| GenBank mRNA | AJ311904.1 | - | 4 | 915 | ||
| AK128729.1 | - | 3 | 268 | |||