Search
Search

ADAMTS15| Gene Symbol: ADAMTS15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139055.3 | NP_620686.1 | 6 | 1754 | ||
| XM_005271419.4 | XP_005271476.1 | 6 | 1038 | |||
| GenBank mRNA | AJ315733.1 | - | 6 | 1754 | ||
| BC109114.1 | - | 6 | 1754 | 40035454 | ||