Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PITX2 SIRNA Assays ›
          Gene Symbol:
          PITX2
          Gene Name:
          paired like homeodomain 2
          Gene Aliases:
          ARP1, Brx1, IDG2, IGDS, IGDS2, IHG2, IRID2, Otlx2, PTX2, RGS, RIEG, RIEG1, RS
          Chromosome Location:
          Chr.4: 110617423 - 110642123 on Build GRCh38
          UniGene ID:
          Hs.643588
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 114053

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PITX2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000325.5 NP_000316.2 3 1758
          NM_001204397.1 NP_001191326.1 7 2922
          NM_001204398.1 NP_001191327.1 5 1314
          NM_001204399.1 NP_001191328.1 4 1176
          NM_153426.2 NP_700475.1 6 2688
          NM_153427.2 NP_700476.1 5 2550
          XM_011532027.2 XP_011530329.1 4 2426
          GenBank mRNA AF048720.1 - 4 1211
          AF048721.1 - 5 1349
          AF048722.1 - 3 1713
          AK127829.1 - 7 2760
          AK291591.1 - 4 1204
          BC013998.2 - 3 1757 3937807
          BC106010.1 - 4 1186 3914727
          U69961.1 - 5 1544

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          patterning of blood vessels
          vasculogenesis
          in utero embryonic development
          neuron migration
          extraocular skeletal muscle development
          atrioventricular valve development
          cardiac neural crest cell migration involved in outflow tract morphogenesis
          pulmonary myocardium development
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          determination of left/right symmetry
          myoblast fusion
          male gonad development
          female gonad development
          response to hormone
          Wnt signaling pathway
          subthalamic nucleus development
          hypothalamus cell migration
          regulation of cell migration
          embryonic camera-type eye development
          response to vitamin A
          embryonic hindlimb morphogenesis
          hair cell differentiation
          vascular smooth muscle cell differentiation
          deltoid tuberosity development
          odontogenesis of dentin-containing tooth
          odontogenesis
          camera-type eye development
          positive regulation of DNA binding
          positive regulation of transcription from RNA polymerase II promoter
          spleen development
          embryonic digestive tract morphogenesis
          atrial cardiac muscle tissue morphogenesis
          ventricular cardiac muscle cell development
          somatotropin secreting cell differentiation
          prolactin secreting cell differentiation
          ventricular septum morphogenesis
          left lung morphogenesis
          pulmonary vein morphogenesis
          superior vena cava morphogenesis
          endodermal digestive tract morphogenesis
          iris morphogenesis
          cell proliferation involved in outflow tract morphogenesis
          left/right axis specification
          positive regulation of myoblast proliferation
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II activating transcription factor binding
          RNA polymerase II transcription coactivator activity
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding
          chromatin DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline