Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002299.3 | NP_002290.2 | 1 | 547 | ||
XM_017004088.1 | XP_016859577.1 | 1 | 567 | |||
GenBank mRNA | X07994.1 | - | 1 | 547 |