Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CADPS SIRNA Assays ›
Gene Symbol:
CADPS
Gene Name:
calcium dependent secretion activator
Gene Aliases:
CADPS1, CAPS, CAPS1, UNC-31
Chromosome Location:
Chr.3: 62398346 - 62875389 on Build GRCh38
UniGene ID:
Hs.654933
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
114363
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CADPS
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_003716.3
NP_003707.2
24
3775
NM_183393.2
NP_899630.1
21
3538
NM_183394.2
NP_899631.1
22
3658
XM_006713378.2
XP_006713441.2
22
3596
XM_011534177.1
XP_011532479.1
26
3989
XM_011534178.1
XP_011532480.1
25
3971
XM_011534180.1
XP_011532482.1
26
3941
XM_011534182.1
XP_011532484.1
25
3920
XM_011534183.1
XP_011532485.1
24
3902
XM_011534189.1
XP_011532491.1
24
3824
XM_011534191.2
XP_011532493.1
23
3445
XM_011534192.1
XP_011532494.1
25
3782
XM_011534193.1
XP_011532495.1
25
3770
XM_011534194.2
XP_011532496.1
25
3427
XM_011534195.1
XP_011532497.1
23
3755
XM_011534196.1
XP_011532498.1
24
3752
XM_011534197.2
XP_011532499.1
22
3701
XM_011534198.2
XP_011532500.1
22
3247
XM_011534199.2
XP_011532501.1
21
3229
XM_011534200.2
XP_011532502.1
21
3229
XM_011534203.1
XP_011532505.1
24
3752
XM_017007358.1
XP_016862847.1
27
4010
XM_017007359.1
XP_016862848.1
27
3998
XM_017007360.1
XP_016862849.1
26
3989
XM_017007361.1
XP_016862850.1
26
3941
XM_017007362.1
XP_016862851.1
26
3929
XM_017007363.1
XP_016862852.1
25
3920
XM_017007364.1
XP_016862853.1
25
3842
XM_017007365.1
XP_016862854.1
25
3830
XM_017007366.1
XP_016862855.1
24
3824
XM_017007367.1
XP_016862856.1
24
3463
XM_017007368.1
XP_016862857.1
23
3445
XM_017007369.1
XP_016862858.1
25
3782
XM_017007370.1
XP_016862859.1
25
3782
XM_017007371.1
XP_016862860.1
24
3773
XM_017007372.1
XP_016862861.1
24
3403
XM_017007373.1
XP_016862862.1
25
3770
XM_017007374.1
XP_016862863.1
24
3761
XM_017007375.1
XP_016862864.1
24
3403
XM_017007376.1
XP_016862865.1
23
3755
XM_017007377.1
XP_016862866.1
23
3394
XM_017007378.1
XP_016862867.1
22
3376
XM_017007379.1
XP_016862868.1
23
3743
XM_017007380.1
XP_016862869.1
24
3686
XM_017007381.1
XP_016862870.1
24
3683
XM_017007382.1
XP_016862871.1
23
3316
XM_017007383.1
XP_016862872.1
23
3665
XM_017007384.1
XP_016862873.1
24
3635
XM_017007385.1
XP_016862874.1
24
3623
XM_017007386.1
XP_016862875.1
23
3614
GenBank mRNA
AB032947.2
-
24
3765
AB593079.1
-
22
3701
AF458662.1
-
21
3488
AK000873.1
-
5
1988
AL833895.1
-
22
2955
BC015754.1
-
1
91
BC072684.1
-
1
56
BC131731.1
-
15
1733
40124561
BX647166.1
-
3
375
U36448.1
-
1
109
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_013219, U16802.
This siRNA also targets the following transcripts in Mouse: NM_001042617, NM_012061, AK133310, AK161919, BC079679, and others.
Panther Classification:
Molecular Function -
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
exocytosis
protein transport
vesicle organization
synaptic vesicle priming
catecholamine secretion
dense core granule exocytosis
protein binding
lipid binding
protein kinase binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0