Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TGIF1 SIRNA Assays ›
Gene Symbol:
TGIF1
Gene Name:
TGFB induced factor homeobox 1
Gene Aliases:
HPE4, TGIF
Chromosome Location:
Chr.18: 3411927 - 3458412 on Build GRCh38
UniGene ID:
Hs.373550
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
115092
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TGIF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001278682.1
NP_001265611.1
2
404
NM_001278684.1
NP_001265613.1
3
761
NM_001278686.1
NP_001265615.1
3
427
NM_003244.3
NP_003235.1
2
494
NM_170695.3
NP_733796.2
2
1138
NM_173207.2
NP_775299.1
2
372
NM_173208.2
NP_775300.1
3
764
NM_173209.2
NP_775301.1
2
580
NM_173210.2
NP_775302.1
2
531
NM_173211.1
NP_775303.1
2
243
NM_174886.2
NP_777480.1
3
561
XM_011525735.2
XP_011524037.1
2
740
XM_017025958.1
XP_016881447.1
3
825
XM_017025959.1
XP_016881448.1
2
610
GenBank mRNA
AF179900.1
-
2
182
AI813434.1
-
3
382
AK026766.1
-
1
1100
AK291112.1
-
2
328
AK309471.1
-
2
489
AU310737.1
-
2
494
BC000814.1
-
2
260
3453337
BC031268.1
-
2
866
5284849
BE296707.1
-
2
256
BE906361.1
-
3
413
BF244060.1
-
2
342
BF446044.1
-
3
376
BM849453.1
-
3
430
BQ437864.1
-
2
492
BU657748.1
-
2
348
BX952985.1
-
2
261
CB161219.1
-
2
404
CB996365.1
-
3
588
CR450333.1
-
2
176
D82120.1
-
2
243
DR007147.1
-
2
584
HY041619.1
-
2
373
X89750.1
-
2
487
Back To Top
More Information
Panther Classification:
Molecular Function -
homeodomain transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
multicellular organism development
response to drug
cellular response to growth factor stimulus
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
protein binding
co-SMAD binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0