Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SALL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127892.1 | NP_001121364.1 | 2 | 246 | ||
NM_002968.2 | NP_002959.2 | 2 | 136 | |||
XM_006721241.3 | XP_006721304.1 | 3 | 211 | |||
XM_011523254.2 | XP_011521556.1 | 3 | 237 | |||
GenBank mRNA | AK295103.1 | - | 2 | 134 | ||
AK307835.1 | - | 2 | 246 | |||
BP229682.1 | - | 2 | 137 | |||
DA318592.1 | - | 2 | 108 | |||
EU668343.1 | - | 2 | 105 | |||
Y18265.1 | - | 2 | 105 |