Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TFAP2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003222.3 | NP_003213.1 | 6 | 1236 | ||
GenBank mRNA | AK301572.1 | - | 6 | 1052 | ||
AK314272.1 | - | 6 | 1232 | |||
BC035664.1 | - | 6 | 1195 | 5723548 | ||
BC051829.1 | - | 6 | 1236 | 6148554 | ||
U85658.1 | - | 6 | 1159 | |||
X95693.1 | - | 6 | 1039 |