Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TEAD2 SIRNA Assays ›
Gene Symbol:
TEAD2
Gene Name:
TEA domain transcription factor 2
Gene Aliases:
ETF, TEAD-2, TEF-4, TEF4
Chromosome Location:
Chr.19: 49340595 - 49362457 on Build GRCh38
UniGene ID:
Hs.515534
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
115445
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TEAD2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256658.1
NP_001243587.1
3, 4
424
NM_001256659.1
NP_001243588.1
3, 4
380
NM_001256660.1
NP_001243589.1
3, 4
420
NM_001256661.1
NP_001243590.1
3, 4
395
NM_001256662.1
NP_001243591.1
2, 3
142
NM_003598.1
NP_003589.1
3, 4
380
XM_005259334.3
XP_005259391.1
2, 3
652
XM_006723424.1
XP_006723487.1
2, 3
652
XM_006723428.2
XP_006723491.1
2, 3
652
XM_011527398.1
XP_011525700.1
2, 3
652
XM_011527399.2
XP_011525701.1
3, 4
375
XM_011527400.2
XP_011525702.1
3, 4
345
XM_011527401.1
XP_011525703.1
3, 4
318
XM_011527402.2
XP_011525704.1
3, 4
320
XM_011527403.1
XP_011525705.1
3, 4
356
XM_011527404.1
XP_011525706.1
2, 3
652
XM_011527405.2
XP_011525707.1
2, 3
652
XM_011527406.2
XP_011525708.1
2, 3
652
GenBank mRNA
AK093236.1
-
2, 3
652
AK290736.1
-
3, 4
377
AK300241.1
-
2, 3
126
AL832240.1
-
3, 4
345
BC007556.1
-
3, 4
380
2967735
BC018803.1
-
3, 4
395
BC051301.1
-
3, 4
374
4843913
BG334109.1
-
3, 4
381
X94440.1
-
2, 3
196
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
hippo signaling
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-hsd7p:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0