Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other JARID2 SIRNA Assays ›
          Gene Symbol:
          JARID2
          Gene Name:
          jumonji and AT-rich interaction domain containing 2
          Gene Aliases:
          JMJ
          Chromosome Location:
          Chr.6: 15245975 - 15522042 on Build GRCh38
          UniGene ID:
          Hs.269059
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 115587

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: JARID2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001267040.1 NP_001253969.1 3 352
          NM_004973.3 NP_004964.2 3 852
          XM_011514579.2 XP_011512881.1 5 996
          XM_011514584.2 XP_011512886.1 1 106
          XM_017010833.1 XP_016866322.1 5 996
          GenBank mRNA AK292861.1 - 3 698
          AK299349.1 - 2 266
          AK302624.1 - 3 347
          AK303610.1 - 3 295
          BC046246.1 - 3 569 4520786
          U57592.1 - 3 531

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: XM_006253776, XM_008771567, XM_003751661.
          This siRNA also targets the following transcripts in Mouse: NM_001205044, NM_001205043, NM_021878, D31967, BC052444, and others.

          Panther Classification:

          Molecular Function -

          histone modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          liver development
          transcription, DNA-templated
          central nervous system development
          negative regulation of cell proliferation
          covalent chromatin modification
          negative regulation of histone methylation
          negative regulation of gene expression, epigenetic
          negative regulation of transcription, DNA-templated
          spleen development
          thymus development
          stem cell differentiation
          positive regulation of histone H3-K9 methylation
          RNA polymerase II regulatory region sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
          chromatin binding
          histone demethylase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0