Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SLC7A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126335.1 | NP_001119807.1 | 5 | 646 | ||
NM_001243036.1 | NP_001229965.1 | 5 | 741 | |||
NM_014270.4 | NP_055085.1 | 5 | 732 | |||
XM_011526402.2 | XP_011524704.1 | 5 | 1836 | |||
XM_017026230.1 | XP_016881719.1 | 3 | 382 | |||
GenBank mRNA | AB033548.1 | - | 5 | 617 | ||
AF141289.1 | - | 5 | 714 | |||
AJ249199.1 | - | 4 | 591 | |||
AK026446.1 | - | 5 | 715 | |||
AK223147.1 | - | 5 | 720 | |||
AK313708.1 | - | 5 | 633 | |||
BC017962.1 | - | 5 | 721 | 4608486 |