Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IKZF2 SIRNA Assays ›
          Gene Symbol:
          IKZF2
          Gene Name:
          IKAROS family zinc finger 2
          Gene Aliases:
          ANF1A2, HELIOS, ZNF1A2, ZNFN1A2
          Chromosome Location:
          Chr.2: 212999684 - 213152753 on Build GRCh38
          UniGene ID:
          Hs.604950
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 116126

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IKZF2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001079526.1 NP_001072994.1 7 857
          NM_016260.2 NP_057344.2 6 788
          XM_005246384.4 XP_005246441.1 6 898
          XM_005246385.3 XP_005246442.1 6 928
          XM_005246386.4 XP_005246443.1 6 779
          XM_011510802.2 XP_011509104.1 7 970
          XM_011510803.2 XP_011509105.1 7 819
          XM_011510804.2 XP_011509106.1 7 742
          XM_011510805.2 XP_011509107.1 7 831
          XM_011510807.2 XP_011509109.1 7 1006
          XM_011510808.2 XP_011509110.1 7 856
          XM_011510809.2 XP_011509111.1 7 868
          XM_011510810.2 XP_011509112.1 7 782
          XM_011510811.2 XP_011509113.1 6 903
          XM_011510812.2 XP_011509114.1 6 764
          XM_011510814.2 XP_011509116.1 6 1262
          XM_011510815.2 XP_011509117.1 6 710
          XM_011510816.2 XP_011509118.1 4 711
          XM_011510817.2 XP_011509119.1 6 790
          XM_011510818.2 XP_011509120.1 4 6365
          XM_011510819.2 XP_011509121.1 6 779
          XM_017003589.1 XP_016859078.1 6 850
          XM_017003590.1 XP_016859079.1 6 820
          XM_017003591.1 XP_016859080.1 4 1253
          XM_017003592.1 XP_016859081.1 6 711
          GenBank mRNA AF130863.1 - 6 788
          AK301503.1 - 6 784
          AY587062.1 - 7 772
          AY587063.1 - 7 734
          AY587064.1 - 5 619
          AY587065.1 - 6 625
          AY587066.1 - 5 541
          AY587067.1 - 5 436
          AY587068.1 - 5 619
          AY587069.1 - 5 541
          AY587070.1 - 3 184
          AY587073.1 - 6 798
          BC028936.1 - 7 857 4540576

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          biological_process
          positive regulation of transcription from RNA polymerase II promoter
          molecular_function
          transcription factor activity, sequence-specific DNA binding
          protein binding
          protein homodimerization activity
          transcription regulatory region DNA binding
          metal ion binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-hqkpn:80/100.66.73.1:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0