Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZNF317 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190791.1 | NP_001177720.1 | 6 | 858 | ||
NM_020933.4 | NP_065984.3 | 7 | 954 | |||
NR_102435.1 | 7 | 1100 | ||||
GenBank mRNA | AB046808.1 | - | 6 | 731 | ||
AF148135.1 | - | 6 | 841 | |||
AF275255.1 | - | 7 | 937 | |||
AF307096.1 | - | 4 | 1075 | |||
AF307097.1 | - | 5 | 1173 | |||
AK056570.1 | - | 7 | 937 | |||
AK304871.1 | - | 6 | 669 | |||
AK314560.1 | - | 7 | 937 | |||
AL834126.2 | - | 5 | 2029 | |||
BC078154.1 | - | 7 | 924 | 6060418 |