Search
Search

PCGF1| Gene Symbol: PCGF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032673.2 | NP_116062.2 | 3 | 313 | ||
| GenBank mRNA | AF087884.1 | - | 2 | 419 | ||
| AK125742.1 | - | 3 | 1199 | |||
| BC004952.1 | - | 3 | 278 | 3621400 | ||
| BU602498.1 | - | 3 | 316 | |||