Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PXN SIRNA Assays ›
          Gene Symbol:
          PXN
          Gene Name:
          paxillin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 120210439 - 120265771 on Build GRCh38
          UniGene ID:
          Hs.446336
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 117087

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PXN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001080855.2 NP_001074324.1 12 1875
          NM_001243756.1 NP_001230685.1 11 1917
          NM_002859.3 NP_002850.2 11 1773
          NM_025157.4 NP_079433.3 11 1897
          XM_005253917.3 XP_005253974.1 7 1272
          XM_006719532.1 XP_006719595.1 15 3458
          XM_006719534.1 XP_006719597.1 15 3613
          XM_006719535.3 XP_006719598.1 16 3541
          XM_006719536.2 XP_006719599.1 15 3430
          XM_011538622.1 XP_011536924.1 16 3540
          XM_011538623.1 XP_011536925.1 16 3727
          XM_017019726.1 XP_016875215.1 15 4026
          XM_017019727.1 XP_016875216.1 15 3826
          XM_017019728.1 XP_016875217.1 15 3817
          XM_017019729.1 XP_016875218.1 15 3882
          XM_017019731.1 XP_016875220.1 16 3911
          XM_017019733.1 XP_016875222.1 13 3441
          XM_017019734.1 XP_016875223.1 15 3338
          XM_017019735.1 XP_016875224.1 12 3195
          XM_017019736.1 XP_016875225.1 13 2865
          XM_017019737.1 XP_016875226.1 13 2721
          XM_017019738.1 XP_016875227.1 12 2556
          XM_017019739.1 XP_016875228.1 11 2454
          XM_017019740.1 XP_016875229.1 12 2412
          XM_017019741.1 XP_016875230.1 12 2203
          XM_017019742.1 XP_016875231.1 11 2310
          XM_017019743.1 XP_016875232.1 12 2011
          GenBank mRNA AB209034.1 - 14 3551
          AK122964.1 - 12 2203
          AK128712.1 - 9 2223
          AK299486.1 - 9 1204
          AK314204.1 - 11 1732
          BC136787.1 - 11 1707 9020777
          BC136794.1 - 11 1707 9020784
          BC144410.1 - 12 1809 9052931
          BM742662.1 - 1 106
          BX648777.1 - 11 1891
          D86862.1 - 11 1733
          D86863.1 - 10 1775
          U14588.1 - 11 1705

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          muscle contraction
          cell adhesion
          cell-matrix adhesion
          signal transduction
          signal complex assembly
          epidermal growth factor receptor signaling pathway
          transforming growth factor beta receptor signaling pathway
          cellular response to reactive oxygen species
          vascular endothelial growth factor receptor signaling pathway
          growth hormone receptor signaling pathway
          integrin binding
          protein binding
          beta-catenin binding
          zinc ion binding
          vinculin binding
          protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-49ts5:80/100.66.72.225:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0