Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PXN SIRNA Assays ›
          Gene Symbol:
          PXN
          Gene Name:
          paxillin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 120210439 - 120265771 on Build GRCh38
          UniGene ID:
          Hs.446336
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 117088

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PXN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001080855.2 NP_001074324.1 12 2566
          NM_001243756.1 NP_001230685.1 11 2608
          NM_002859.3 NP_002850.2 11 2464
          NM_025157.4 NP_079433.3 11 2588
          XM_005253917.3 XP_005253974.1 7 1963
          XM_006719532.1 XP_006719595.1 15 4149
          XM_006719534.1 XP_006719597.1 15 4304
          XM_006719535.3 XP_006719598.1 16 4232
          XM_006719536.2 XP_006719599.1 15 4121
          XM_011538622.1 XP_011536924.1 16 4231
          XM_011538623.1 XP_011536925.1 16 4418
          XM_017019726.1 XP_016875215.1 15 4717
          XM_017019727.1 XP_016875216.1 15 4517
          XM_017019728.1 XP_016875217.1 15 4508
          XM_017019729.1 XP_016875218.1 15 4573
          XM_017019731.1 XP_016875220.1 16 4602
          XM_017019733.1 XP_016875222.1 13 4132
          XM_017019734.1 XP_016875223.1 15 4029
          XM_017019735.1 XP_016875224.1 12 3886
          XM_017019736.1 XP_016875225.1 13 3556
          XM_017019737.1 XP_016875226.1 13 3412
          XM_017019738.1 XP_016875227.1 12 3247
          XM_017019739.1 XP_016875228.1 11 3145
          XM_017019740.1 XP_016875229.1 12 3103
          XM_017019741.1 XP_016875230.1 12 2894
          XM_017019742.1 XP_016875231.1 11 3001
          XM_017019743.1 XP_016875232.1 12 2702
          GenBank mRNA AB209034.1 - 14 4242
          AK122964.1 - 12 2894
          AK128712.1 - 9 2914
          AK298057.1 - 4 807
          BC003647.2 - 4 738
          BC006840.1 - 1 294
          BC010498.1 - 1 502
          BC015217.1 - 1 369
          BC136787.1 - 11 2398 9020777
          BC136794.1 - 11 2398 9020784
          BC144410.1 - 12 2500 9052931
          BX648777.1 - 11 2581
          U14588.1 - 11 2396

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          muscle contraction
          cell adhesion
          cell-matrix adhesion
          signal transduction
          signal complex assembly
          epidermal growth factor receptor signaling pathway
          transforming growth factor beta receptor signaling pathway
          cellular response to reactive oxygen species
          vascular endothelial growth factor receptor signaling pathway
          growth hormone receptor signaling pathway
          integrin binding
          protein binding
          beta-catenin binding
          zinc ion binding
          vinculin binding
          protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0