Search
Search

SUV39H1| Gene Symbol: SUV39H1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282166.1 | NP_001269095.1 | 3 | 645 | ||
| NM_003173.3 | NP_003164.1 | 3 | 398 | |||
| GenBank mRNA | AF019968.1 | - | 3 | 391 | ||
| AK223071.1 | - | 3 | 370 | |||
| AK297566.1 | - | 677 | ||||
| AK298233.1 | - | 3 | 373 | |||
| AK299900.1 | - | 3 | 645 | |||
| AK312547.1 | - | 3 | 370 | |||
| BC006238.2 | - | 3 | 389 | 3945632 | ||
| CR541746.1 | - | 3 | 346 | |||
| L08238.1 | - | 2 | 488 | |||