Search
Search

FDX1| Gene Symbol: FDX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004109.4 | NP_004100.1 | 4 | 1190 | ||
| GenBank mRNA | BC010284.1 | - | 4 | 1159 | 3048743 | |
| BC017063.1 | - | 4 | 1140 | 3883924 | ||
| J03548.1 | - | 4 | 1134 | |||
| M34788.1 | - | 4 | 1024 | |||