Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WNK1 SIRNA Assays ›
          Gene Symbol:
          WNK1
          Gene Name:
          WNK lysine deficient protein kinase 1
          Gene Aliases:
          HSAN2, HSN2, KDP, PPP1R167, PRKWNK1, PSK, p65
          Chromosome Location:
          Chr.12: 752552 - 911452 on Build GRCh38
          UniGene ID:
          Hs.744906
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 1174

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price 6,301.00
          Your Price
          Online offer:
          6301.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: WNK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001184985.1 NP_001171914.1 1 1254
          NM_014823.2 NP_055638.2 1 1254
          NM_018979.3 NP_061852.3 1 1254
          NM_213655.4 NP_998820.3 1 1254
          XM_006719003.2 XP_006719066.1 1 1579
          XM_011520997.2 XP_011519299.1 1 1625
          XM_011520998.2 XP_011519300.1 1 1579
          XM_011520999.2 XP_011519301.1 1 1579
          XM_011521000.2 XP_011519302.1 1 1579
          XM_011521001.2 XP_011519303.1 1 1579
          XM_011521002.2 XP_011519304.1 1 1579
          XM_011521003.2 XP_011519305.1 1 1579
          XM_011521004.2 XP_011519306.1 1 1579
          XM_011521006.2 XP_011519308.1 1 1579
          XM_011521007.2 XP_011519309.1 1 1579
          XM_011521008.2 XP_011519310.1 1 1579
          XM_011521009.2 XP_011519311.1 1 1579
          XM_017019834.1 XP_016875323.1 1 1579
          XM_017019835.1 XP_016875324.1 1 1579
          XM_017019836.1 XP_016875325.1 1 1579
          XM_017019837.1 XP_016875326.1 1 1579
          XM_017019838.1 XP_016875327.1 1 1579
          XM_017019839.1 XP_016875328.1 1 1579
          GenBank mRNA AB002342.2 - 1 409
          AF061944.1 - 1 1254
          AJ296290.1 - 1 611
          JQ358908.1 - 1 611

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001199095, NM_001002823, NM_053794, XM_008763213, XM_008763201, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001185020, NM_001199083, NM_001185021, NM_001199084, NM_198703, and others.

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of systemic arterial blood pressure
          protein phosphorylation
          ion transport
          negative regulation of phosphatase activity
          peptidyl-threonine phosphorylation
          positive regulation of ion transmembrane transporter activity
          intracellular signal transduction
          protein autophosphorylation
          neuron development
          regulation of cellular process
          negative regulation of protein serine/threonine kinase activity
          negative regulation of pancreatic juice secretion
          regulation of ion homeostasis
          magnesium ion binding
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          chloride channel inhibitor activity
          potassium channel inhibitor activity
          protein kinase binding
          phosphatase binding
          protein serine/threonine kinase inhibitor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0