Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SLC22A15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018420.2 | NP_060890.2 | 11 | 1660 | ||
GenBank mRNA | AK125072.1 | - | 9 | 1228 | ||
AL353933.1 | - | 3 | 361 | |||
AY145501.1 | - | 11 | 1530 | |||
BC026358.1 | - | 11 | 1659 | 4814675 | ||
KU178698.1 | - | 11 | 1494 | |||
KU178699.1 | - | 11 | 1752 | |||
KU178700.1 | - | 10 | 1421 | |||
KU178701.1 | - | 11 | 1418 | |||
KU178702.1 | - | 11 | 1417 | |||
KU178703.1 | - | 10 | 1281 |