Search
Search

PYROXD1| Gene Symbol: PYROXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024854.3 | NP_079130.2 | 4 | 481 | ||
| XM_006719153.3 | XP_006719216.1 | 4 | 455 | |||
| GenBank mRNA | AK025681.1 | - | 4 | 383 | ||
| AK125461.1 | - | 4 | 894 | |||
| AK293820.1 | - | 4 | 470 | |||
| AK307710.1 | - | 4 | 380 | |||
| AL832441.1 | - | 4 | 408 | |||
| BC021662.1 | - | 4 | 382 | 3826938 | ||
| DB041552.1 | - | 4 | 481 | |||