Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC23A1 SIRNA Assays ›
          Gene Symbol:
          SLC23A1
          Gene Name:
          solute carrier family 23 member 1
          Gene Aliases:
          SLC23A2, SVCT1, YSPL3
          Chromosome Location:
          Chr.5: 139367196 - 139384553 on Build GRCh38
          UniGene ID:
          Hs.643467
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 117768

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SLC23A1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005847.4 NP_005838.3 3 327
          NM_152685.3 NP_689898.2 3 327
          XM_005272148.3 XP_005272205.3 4 452
          XM_005272149.3 XP_005272206.3 4 452
          XM_006714741.2 XP_006714804.2 4 452
          XM_011543765.2 XP_011542067.1 4 452
          XM_011543766.1 XP_011542068.1 4 452
          XM_011543767.1 XP_011542069.1 4 452
          XM_011543768.1 XP_011542070.1 3 278
          XM_011543769.1 XP_011542071.1 4 452
          GenBank mRNA AF098277.1 - 3 277
          AF170911.1 - 3 261
          AJ250807.1 - 3 300
          AJ269477.1 - 3 230
          AK092655.1 - 2 139
          AK290789.1 - 3 248
          AK298337.1 - 3 287
          AK300935.1 - 3 274
          BC019225.1 - 3 292
          BC050261.1 - 3 269 5188430
          DA385445.1 - 3 328

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleobase-containing compound metabolic process
          sodium ion transport
          brain development
          response to toxic substance
          nucleobase transport
          L-ascorbic acid transport
          L-ascorbic acid metabolic process
          lung development
          vitamin transmembrane transport
          sodium ion transmembrane transport
          dehydroascorbic acid transport
          transepithelial L-ascorbic acid transport
          protein binding
          L-ascorbate:sodium symporter activity
          sodium ion transmembrane transporter activity
          nucleobase transmembrane transporter activity
          L-ascorbic acid transporter activity
          dehydroascorbic acid transporter activity
          sodium-dependent L-ascorbate transmembrane transporter activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-kzg96:80/100.66.78.128:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0