Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other POLM SIRNA Assays ›
Gene Symbol:
POLM
Gene Name:
DNA polymerase mu
Gene Aliases:
Pol Mu, Tdt-N
Chromosome Location:
Chr.7: 44072062 - 44082540 on Build GRCh38
UniGene ID:
Hs.596982
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
117949
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: POLM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001284330.1
NP_001271259.1
2
306
NM_001284331.1
NP_001271260.1
2
306
NM_013284.3
NP_037416.1
2
306
NR_104299.1
2
306
XM_005249711.2
XP_005249768.1
2
277
XM_006715692.2
XP_006715755.1
2
277
XM_006715696.2
XP_006715759.1
2
277
XM_011515275.1
XP_011513577.1
2
277
XM_011515278.1
XP_011513580.1
2
277
XM_011515279.1
XP_011513581.1
2
277
XM_011515286.2
XP_011513588.1
2
277
XM_011515287.2
XP_011513589.1
2
277
XM_017011998.1
XP_016867487.1
2
277
XM_017011999.1
XP_016867488.1
2
277
XM_017012000.1
XP_016867489.1
2
277
XM_017012001.1
XP_016867490.1
2
277
XM_017012002.1
XP_016867491.1
2
259
XM_017012003.1
XP_016867492.1
2
277
XM_017012004.1
XP_016867493.1
2
277
XM_017012005.1
XP_016867494.1
2
277
GenBank mRNA
AF176097.1
-
2
246
AJ131891.2
-
2
249
AK023002.1
-
2
281
AK092903.1
-
2
256
AK291638.1
-
2
250
AK294449.1
-
2
277
AK307634.1
-
2
249
AU309799.1
-
2
306
BC026306.1
-
2
277
4813399
BC049202.1
-
2
251
5246144
BC062590.1
-
2
230
5215523
DC407437.1
-
2
296
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-directed DNA polymerase
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair via nonhomologous end joining
DNA recombination
somatic hypermutation of immunoglobulin genes
B cell differentiation
DNA biosynthetic process
DNA binding
DNA-directed DNA polymerase activity
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-5btgp:80/100.66.77.235:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0