Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ABCG2 SIRNA Assays ›
          Gene Symbol:
          ABCG2
          Gene Name:
          ATP binding cassette subfamily G member 2 (Junior blood group)
          Gene Aliases:
          ABC15, ABCP, BCRP, BCRP1, BMDP, CD338, CDw338, EST157481, GOUT1, MRX, MXR, MXR-1, MXR1, UAQTL1
          Chromosome Location:
          Chr.4: 88090264 - 88231417 on Build GRCh38
          UniGene ID:
          Hs.480218
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 118155

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ABCG2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001257386.1 NP_001244315.1 3, 4 609
          NM_004827.2 NP_004818.2 3, 4 754
          XM_005263354.3 XP_005263411.1 3, 4 597
          XM_005263355.3 XP_005263412.1 3, 4 704
          XM_005263356.3 XP_005263413.1 3, 4 480
          XM_011532420.2 XP_011530722.1 5, 6 868
          XM_017008852.1 XP_016864341.1 3, 4 597
          XM_017008853.1 XP_016864342.1 3, 4 394
          GenBank mRNA AB051855.1 - 3, 4 305
          AB056867.1 - 2, 3 261
          AF098951.2 - 3, 4 499
          AF103796.1 - 3, 4 465
          AF463519.1 - 2, 3 269
          AK290000.1 - 3, 4 325
          AY017168.1 - 3, 4 454
          AY289766.1 - 3, 4 461
          AY333755.1 - 2, 3 270
          AY333756.1 - 2, 3 270
          BC021281.2 - 3, 4 754 4908836
          BC092408.1 - 3, 4 609 6181905
          DQ459562.1 - 3, 4 383
          DQ459564.1 - 3, 4 374
          DQ459567.1 - 3, 4 374
          DQ459568.1 - 3, 4 375
          DQ459570.1 - 3, 4 466
          DQ459571.1 - 3, 4 466
          DQ459573.1 - 3, 4 356
          DQ459574.1 - 3, 4 315
          DQ459575.1 - 3, 4 309
          DQ459576.1 - 3, 4 381
          DQ459577.1 - 3, 4 379
          DQ459578.1 - 3, 4 375
          DQ459579.1 - 3, 4 375
          DQ459580.1 - 3, 4 332
          DQ459582.1 - 3, 4 566
          DQ459583.1 - 3, 4 518
          DQ459584.1 - 3, 4 518
          DQ459585.1 - 3, 4 517
          DQ459586.1 - 3, 4 515
          DQ459587.1 - 3, 4 508
          DQ459588.1 - 3, 4 411
          DQ459589.1 - 3, 4 349
          DQ459591.1 - 3, 4 371
          DQ459592.1 - 3, 4 359
          DQ459593.1 - 3, 4 399
          DQ459594.1 - 3, 4 399
          DQ459595.1 - 3, 4 376
          DQ459596.1 - 3, 4 374
          DQ459597.1 - 3, 4 494
          DQ459598.1 - 3, 4 466
          DQ459600.1 - 3, 4 359
          DQ459601.1 - 3, 4 359
          DQ459602.1 - 3, 4 359
          DQ459603.1 - 3, 4 312
          DQ459604.1 - 3, 4 375
          DQ459605.1 - 3, 4 339
          DQ459606.1 - 3, 4 331
          DQ459607.1 - 2, 3 275
          DQ459608.1 - 2, 3 200
          DQ459609.1 - 3, 4 315
          DQ459610.1 - 3, 4 361
          DQ459611.1 - 3, 4 359
          DQ459612.1 - 3, 4 355
          DQ459613.1 - 3, 4 355
          DQ459614.1 - 3, 4 334
          HQ386990.1 - 3, 4 359
          HQ386991.1 - 3, 4 359
          HQ386992.1 - 3, 4 359
          HQ386993.1 - 3, 4 359
          HQ386994.1 - 3, 4 359
          HQ386995.1 - 3, 4 359
          HQ386996.1 - 3, 4 359
          HQ386997.1 - 3, 4 359
          HQ386998.1 - 3, 4 518
          HQ386999.1 - 3, 4 518
          HQ387000.1 - 3, 4 518
          HQ387001.1 - 3, 4 518
          HQ387002.1 - 3, 4 518
          HQ387003.1 - 3, 4 518
          HQ387004.1 - 3, 4 518
          HQ387005.1 - 3, 4 518
          HQ387006.1 - 3, 4 566
          HQ387007.1 - 3, 4 375

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ATP-binding cassette (ABC) transporter primary active transporter transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          drug transmembrane transport
          cellular iron ion homeostasis
          response to iron ion
          heme transport
          response to drug
          xenobiotic transport
          urate metabolic process
          drug export
          response to folic acid
          embryonic process involved in female pregnancy
          cellular response to dexamethasone stimulus
          transporter activity
          protein binding
          ATP binding
          xenobiotic-transporting ATPase activity
          heme transporter activity
          ATPase activity, coupled to transmembrane movement of substances
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-4p4qf:80/100.66.72.111:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0