Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ABCG1 SIRNA Assays ›
          Gene Symbol:
          ABCG1
          Gene Name:
          ATP binding cassette subfamily G member 1
          Gene Aliases:
          ABC8, WHITE1
          Chromosome Location:
          Chr.21: 42199689 - 42304389 on Build GRCh38
          UniGene ID:
          Hs.124649
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 118196

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ABCG1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004915.3 NP_004906.3 15 2040
          NM_016818.2 NP_058198.2 15 2004
          NM_207174.1 NP_997057.1 15 2005
          NM_207627.1 NP_997510.1 16 2164
          NM_207628.1 NP_997511.1 17 2082
          NM_207629.1 NP_997512.1 15 1968
          XM_011529806.1 XP_011528108.1 15 2118
          XM_011529807.2 XP_011528109.1 15 2140
          GenBank mRNA AF038175.1 - 5 596
          AF323660.1 - 16 2164
          AF323661.1 - 17 2082
          AF323662.1 - 15 1968
          AF323663.1 - 15 1927
          AF323664.1 - 15 2005
          AK092959.1 - 11 1879
          AK292152.1 - 15 2014
          AK298491.1 - 15 2118
          AL833689.1 - 1 720
          AY048757.1 - 16 2520
          BC029158.1 - 15 2004 5198758
          X91249.1 - 15 1952

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ATP-binding cassette (ABC) transporter primary active transporter transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          cholesterol metabolic process
          detection of hormone stimulus
          response to organic substance
          negative regulation of macrophage derived foam cell differentiation
          regulation of cholesterol esterification
          positive regulation of cholesterol efflux
          negative regulation of cholesterol storage
          intracellular cholesterol transport
          cholesterol efflux
          phospholipid efflux
          response to lipid
          low-density lipoprotein particle remodeling
          high-density lipoprotein particle remodeling
          glycoprotein transport
          lipoprotein metabolic process
          cholesterol homeostasis
          amyloid precursor protein catabolic process
          reverse cholesterol transport
          positive regulation of cholesterol biosynthetic process
          transmembrane transport
          phospholipid homeostasis
          response to high density lipoprotein particle
          toxin transport
          ATP binding
          phospholipid binding
          phospholipid transporter activity
          cholesterol binding
          cholesterol transporter activity
          toxin transporter activity
          sterol-transporting ATPase activity
          glycoprotein transporter activity
          protein homodimerization activity
          ADP binding
          protein heterodimerization activity
          protein dimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0