Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PIP5K1A SIRNA Assays ›
Gene Symbol:
PIP5K1A
Gene Name:
phosphatidylinositol-4-phosphate 5-kinase type 1 alpha
Gene Aliases:
-
Chromosome Location:
Chr.1: 151198003 - 151249536 on Build GRCh38
UniGene ID:
Hs.655131
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
118246
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PIP5K1A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135636.1
NP_001129108.1
13
1986
NM_001135637.1
NP_001129109.1
13
1920
NM_001135638.1
NP_001129110.1
15
2106
NM_003557.2
NP_003548.1
14
2067
XM_005245525.4
XP_005245582.1
16
2347
XM_005245527.3
XP_005245584.1
15
2096
XM_005245528.4
XP_005245585.1
15
2311
XM_005245530.4
XP_005245587.1
15
1669
XM_005245531.3
XP_005245588.1
14
2063
XM_005245532.3
XP_005245589.1
14
1996
XM_006711563.3
XP_006711626.1
16
2353
XM_006711564.3
XP_006711627.1
15
2065
XM_006711567.3
XP_006711630.1
16
1653
XM_006711568.3
XP_006711631.2
15
1684
XM_011510043.2
XP_011508345.1
14
1648
XM_011510045.2
XP_011508347.1
15
1666
XM_011510046.2
XP_011508348.1
14
2141
XM_011510048.2
XP_011508350.1
16
1663
XM_011510049.2
XP_011508351.1
16
1650
XM_017002439.1
XP_016857928.1
12
1786
XM_017002440.1
XP_016857929.1
12
1783
XM_017002441.1
XP_016857930.1
16
1669
GenBank mRNA
AK291015.1
-
14
2010
AK295691.1
-
13
1979
AK302268.1
-
6
505
BC007833.2
-
13
1912
4098514
U78575.1
-
14
2015
U78576.1
-
15
2075
U78577.1
-
13
1869
U78578.1
-
7
591
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
Gene Ontology Categories:
Function(s)
Process(es)
glycerophospholipid metabolic process
phosphatidylinositol biosynthetic process
phagocytosis
signal transduction
phospholipid biosynthetic process
fibroblast migration
regulation of phosphatidylinositol 3-kinase signaling
cell migration
keratinocyte differentiation
actin cytoskeleton reorganization
phosphatidylinositol phosphorylation
focal adhesion assembly
cell chemotaxis
protein targeting to plasma membrane
activation of GTPase activity
ruffle assembly
1-phosphatidylinositol-3-phosphate 5-kinase activity
protein binding
ATP binding
1-phosphatidylinositol-4-phosphate 5-kinase activity
kinase binding
1-phosphatidylinositol-5-kinase activity
1-phosphatidylinositol-3-phosphate 4-kinase activity
phosphatidylinositol-3,4-bisphosphate 5-kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-wpngx:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0