Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001372.3 | NP_001363.2 | 57 | 11146 | ||
NM_004662.2 | NP_004653.2 | 3 | 543 | |||
XM_011523703.1 | XP_011522005.1 | 57 | 11110 | |||
XM_017024292.1 | XP_016879781.1 | 56 | 10993 | |||
XM_017024293.1 | XP_016879782.1 | 46 | 9054 | |||
GenBank mRNA | AB002355.2 | - | 38 | 6824 | ||
AB290158.1 | - | 45 | 8963 | |||
AF257737.1 | - | 57 | 11136 | |||
AJ404468.1 | - | 57 | 11146 | |||
AK302009.1 | - | 2 | 359 | |||
BC128261.1 | - | 1 | 83 | |||
BC128421.1 | - | 1 | 83 | |||
BC153883.1 | - | 38 | 6824 | |||
X99947.1 | - | 3 | 543 |