Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DYNC1H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001376.4 | NP_001367.2 | 4 | 850 | ||
GenBank mRNA | AB002323.3 | - | 4 | 722 | ||
AB290157.1 | - | 4 | 850 | |||
AK299878.1 | - | 4 | 847 |