Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MYO18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318245.1 | NP_001305174.1 | 4 | 1325 | ||
NM_032608.6 | NP_115997.5 | 4 | 1325 | |||
XM_011530458.2 | XP_011528760.1 | 5 | 1461 | |||
XM_011530459.2 | XP_011528761.1 | 5 | 1432 | |||
XM_011530460.2 | XP_011528762.1 | 4 | 1305 | |||
XM_011530461.2 | XP_011528763.1 | 5 | 1461 | |||
XM_011530464.2 | XP_011528766.1 | 5 | 1461 | |||
XM_011530465.2 | XP_011528767.1 | 4 | 1218 | |||
XM_011530466.2 | XP_011528768.1 | 5 | 1462 | |||
XM_017029012.1 | XP_016884501.1 | 5 | 1461 | |||
XM_017029013.1 | XP_016884502.1 | 5 | 1509 | |||
XM_017029014.1 | XP_016884503.1 | 5 | 1461 | |||
XM_017029015.1 | XP_016884504.1 | 4 | 1335 | |||
XM_017029017.1 | XP_016884506.1 | 4 | 1334 | |||
GenBank mRNA | AB075376.1 | - | 4 | 1325 | ||
AJ310931.2 | - | 4 | 1316 | |||
AY077700.1 | - | 4 | 1235 | |||
BC144596.1 | - | 3 | 1125 | |||
BC144597.1 | - | 3 | 1125 | 9053127 | ||
BC150626.1 | - | 3 | 1125 | 9056996 |