Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPOX SIRNA Assays ›
Gene Symbol:
PPOX
Gene Name:
protoporphyrinogen oxidase
Gene Aliases:
PPO, V290M, VP
Chromosome Location:
Chr.1: 161165728 - 161178277 on Build GRCh38
UniGene ID:
Hs.517373
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
118551
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPOX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000309.3
NP_000300.1
8
1080
NM_001122764.1
NP_001116236.1
8
1141
XM_005245291.3
XP_005245348.2
8
1119
XM_005245295.3
XP_005245352.2
7
967
XM_006711402.2
XP_006711465.2
8
1086
XM_006711403.2
XP_006711466.2
8
1075
XM_006711404.3
XP_006711467.2
8
1023
XM_006711406.2
XP_006711469.2
8
996
XM_011509663.2
XP_011507965.1
8
1206
XM_011509664.1
XP_011507966.1
8
977
XM_011509665.2
XP_011507967.1
8
1204
XM_011509666.2
XP_011507968.1
8
1211
XM_011509667.2
XP_011507969.1
8
1147
XM_011509668.2
XP_011507970.1
8
947
XM_011509669.1
XP_011507971.1
8
960
XM_011509670.2
XP_011507972.1
7
1095
XM_011509671.1
XP_011507973.1
8
977
XM_011509672.2
XP_011507974.1
7
1086
XM_011509673.2
XP_011507975.1
6
943
XM_011509675.2
XP_011507977.1
7
1028
XM_011509676.1
XP_011507978.1
7
1291
XM_011509677.1
XP_011507979.1
7
1224
XM_011509678.1
XP_011507980.1
8
1184
XM_011509679.1
XP_011507981.1
8
992
XM_011509681.1
XP_011507983.1
7
909
XM_011509682.1
XP_011507984.1
7
919
XM_017001559.1
XP_016857048.1
8
1203
XM_017001560.1
XP_016857049.1
8
1119
XM_017001561.1
XP_016857050.1
7
1612
XM_017001562.1
XP_016857051.1
8
1065
XM_017001563.1
XP_016857052.1
8
1163
XM_017001564.1
XP_016857053.1
4
681
XM_017001565.1
XP_016857054.1
8
1065
XM_017001566.1
XP_016857055.1
8
1174
XM_017001567.1
XP_016857056.1
7
967
XM_017001568.1
XP_016857057.1
8
1155
XM_017001569.1
XP_016857058.1
7
909
XM_017001570.1
XP_016857059.1
5
710
GenBank mRNA
AK094855.1
-
7
913
AK302298.1
-
7
951
BC002357.2
-
8
1061
2821983
D38537.1
-
8
1128
U26446.1
-
8
1042
Back To Top
More Information
Panther Classification:
Molecular Function -
oxidase
Gene Ontology Categories:
Function(s)
Process(es)
porphyrin-containing compound biosynthetic process
protoporphyrinogen IX biosynthetic process
heme biosynthetic process
response to drug
oxidation-reduction process
oxygen-dependent protoporphyrinogen oxidase activity
flavin adenine dinucleotide binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0