Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TIMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003256.3 | NP_003247.1 | 5 | 1208 | ||
GenBank mRNA | BC010553.1 | - | 5 | 1208 | 4154090 | |
U76456.1 | - | 5 | 756 |