Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SERPINB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307923.1 | NP_001294852.1 | 8 | 2294 | ||
NM_012397.3 | NP_036529.1 | 8 | 2267 | |||
XM_005266707.3 | XP_005266764.1 | 9 | 2571 | |||
XM_011526029.2 | XP_011524331.1 | 8 | 2218 | |||
GenBank mRNA | AJ001696.2 | - | 8 | 2205 | ||
AJ001697.2 | - | 8 | 2173 | |||
AJ001698.1 | - | 9 | 2352 | |||
AL050177.1 | - | 1 | 799 |