Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: POLR2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032940.2 | NP_116558.1 | 2, 3 | 217 | ||
GenBank mRNA | AF008443.1 | - | 2, 3 | 131 | ||
AJ224143.1 | - | 2, 3 | 170 | |||
AJ224144.1 | - | 2, 3 | 170 | |||
AK293112.1 | - | 2, 3 | 175 | |||
AK295582.1 | - | 2, 3 | 196 | |||
AK300124.1 | - | 188 | ||||
BC000409.2 | - | 2, 3 | 148 | 2960943 | ||
BC003159.1 | - | 2, 3 | 189 | 2957867 | ||
BC028157.1 | - | 2, 3 | 173 | 5240953 | ||
BI458170.1 | - | 2, 3 | 225 | |||
CR542041.1 | - | 2, 3 | 131 | |||
J05448.1 | - | 2, 3 | 172 |