Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARHGEF2 SIRNA Assays ›
Gene Symbol:
ARHGEF2
Gene Name:
Rho/Rac guanine nucleotide exchange factor 2
Gene Aliases:
GEF, GEF-H1, GEFH1, LFP40, P40
Chromosome Location:
Chr.1: 155946839 - 155991263 on Build GRCh38
UniGene ID:
Hs.743352
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
119230
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARHGEF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001162383.1
NP_001155855.1
7
711
NM_001162384.1
NP_001155856.1
6, 7
708
NM_004723.3
NP_004714.2
6, 7
636
XM_005245587.2
XP_005245644.1
7
764
XM_005245588.2
XP_005245645.1
6, 7
761
XM_005245589.4
XP_005245646.1
7
917
XM_005245590.3
XP_005245647.1
7
639
XM_005245591.3
XP_005245648.1
8
755
XM_005245592.2
XP_005245649.1
7
908
XM_005245593.4
XP_005245650.1
7
639
XM_005245594.2
XP_005245651.1
7
1625
XM_006711622.3
XP_006711685.1
7
1902
XM_006711623.3
XP_006711686.1
6
638
XM_006711624.3
XP_006711687.1
6
665
XM_006711625.3
XP_006711688.1
6
571
XM_006711626.2
XP_006711689.1
6
1557
XM_011510136.2
XP_011508438.2
10, 11
1320
XM_011510137.2
XP_011508439.2
11
1323
XM_011510139.2
XP_011508441.1
11
1323
XM_011510141.2
XP_011508443.1
7
1441
XM_011510143.2
XP_011508445.1
3
187
XM_017002794.1
XP_016858283.1
10, 11
1320
XM_017002795.1
XP_016858284.1
6, 7
655
XM_017002796.1
XP_016858285.1
6, 7
1899
XM_017002797.1
XP_016858286.1
6, 7
639
XM_017002798.1
XP_016858287.1
6
1834
XM_017002799.1
XP_016858288.1
5, 6
635
GenBank mRNA
AB014551.3
-
10, 11
979
AF486838.1
-
6, 7
590
AK295924.1
-
7
908
AK304548.1
-
10
1048
AL512715.1
-
6, 7
621
BC020567.1
-
7
693
4157775
BT007407.1
-
6
512
U72206.1
-
7
695
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
establishment of mitotic spindle orientation
cell morphogenesis
intracellular protein transport
actin filament organization
negative regulation of microtubule depolymerization
mitotic nuclear division
positive regulation of interleukin-6 production
positive regulation of tumor necrosis factor production
regulation of Rho protein signal transduction
intracellular signal transduction
regulation of cell proliferation
positive regulation of apoptotic process
positive regulation of GTPase activity
innate immune response
positive regulation of transcription from RNA polymerase II promoter
positive regulation of peptidyl-tyrosine phosphorylation
negative regulation of neurogenesis
regulation of small GTPase mediated signal transduction
positive regulation of NF-kappaB transcription factor activity
cell division
negative regulation of necroptotic process
cellular response to muramyl dipeptide
cellular response to tumor necrosis factor
cellular hyperosmotic response
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of intrinsic apoptotic signaling pathway in response to osmotic stress
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
protein binding
microtubule binding
transcription factor binding
zinc ion binding
Rho GTPase binding
Rac guanyl-nucleotide exchange factor activity
Rac GTPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-zznkm:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0