Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ARHGEF7 SIRNA Assays ›
          Gene Symbol:
          ARHGEF7
          Gene Name:
          Rho guanine nucleotide exchange factor 7
          Gene Aliases:
          BETA-PIX, COOL-1, COOL1, Nbla10314, P50, P50BP, P85, P85COOL1, P85SPR, PAK3, PIXB
          Chromosome Location:
          Chr.13: 111114619 - 111305734 on Build GRCh38
          UniGene ID:
          Hs.508738
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 119397

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ARHGEF7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001113511.2 NP_001106983.1 9 1894
          NM_001113512.2 NP_001106984.1 7 1744
          NM_001113513.1 NP_001106985.1 7 836
          NM_001320851.1 NP_001307780.1 6 617
          NM_001320852.1 NP_001307781.1 8 1831
          NM_001320853.1 NP_001307782.1 7 742
          NM_001320854.1 NP_001307783.1 3 356
          NM_003899.3 NP_003890.1 7 949
          NM_145735.3 NP_663788.1 8 1831
          XM_006719956.3 XP_006720019.1 8 959
          XM_011521133.2 XP_011519435.1 8 959
          XM_017020814.1 XP_016876303.1 8 959
          XM_017020815.1 XP_016876304.1 6 952
          XM_017020816.1 XP_016876305.1 7 727
          XM_017020817.1 XP_016876306.1 6 952
          XM_017020818.1 XP_016876307.1 8 827
          XM_017020819.1 XP_016876308.1 7 836
          XM_017020820.1 XP_016876309.1 7 959
          XM_017020821.1 XP_016876310.1 7 654
          XM_017020822.1 XP_016876311.1 6 650
          XM_017020823.1 XP_016876312.1 6 1427
          XM_017020824.1 XP_016876313.1 8 827
          XM_017020825.1 XP_016876314.1 6 578
          XM_017020826.1 XP_016876315.1 8 827
          XM_017020827.1 XP_016876316.1 6 651
          XM_017020828.1 XP_016876317.1 3 216
          XM_017020829.1 XP_016876318.1 3 216
          GenBank mRNA AB177848.1 - 8 1832
          AB177849.1 - 5 555
          AK295472.1 - 3 281
          AK300272.1 - 7 729
          AK308897.1 - 3 356
          AL834228.1 - 8 1015
          BC033905.1 - 7 941 5262912
          BC050521.1 - 7 808
          BC060776.1 - 9 1236 5271977
          D63476.1 - 7 925

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          hematopoietic progenitor cell differentiation
          Golgi organization
          signal transduction
          nervous system development
          positive regulation of fibroblast migration
          lamellipodium assembly
          positive regulation of protein binding
          regulation of Rho protein signal transduction
          intracellular signal transduction
          negative regulation of epidermal growth factor receptor signaling pathway
          positive regulation of apoptotic process
          positive regulation of GTPase activity
          ephrin receptor signaling pathway
          focal adhesion assembly
          regulation of small GTPase mediated signal transduction
          positive regulation of growth hormone secretion
          positive regulation of substrate adhesion-dependent cell spreading
          regulation of GTP binding
          positive regulation of lamellipodium morphogenesis
          guanyl-nucleotide exchange factor activity
          Rho guanyl-nucleotide exchange factor activity
          protein binding
          protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-t8hgc:80/100.66.75.150:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0