Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SLC29A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174098.1 | NP_001167569.1 | 6 | 2121 | ||
NM_018344.5 | NP_060814.4 | 6 | 2115 | |||
NR_033413.1 | 5 | 2032 | ||||
NR_033414.1 | 4 | 1805 | ||||
XM_006717910.3 | XP_006717973.1 | 6 | 2507 | |||
XM_017016377.1 | XP_016871866.1 | 5 | 2329 | |||
XM_017016378.1 | XP_016871867.1 | 4 | 2105 | |||
GenBank mRNA | AK002022.1 | - | 5 | 1866 | ||
AX775915.1 | - | 6 | 2115 | |||
AX775917.1 | - | 6 | 2115 | |||
AX775919.1 | - | 6 | 2115 | |||
AX775921.1 | - | 6 | 2115 | |||
AY358686.1 | - | 6 | 2093 | |||
BC000223.1 | - | 3 | 1490 | |||
BC041575.1 | - | 5 | 2003 |