Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ATP2B2 SIRNA Assays ›
          Gene Symbol:
          ATP2B2
          Gene Name:
          ATPase plasma membrane Ca2+ transporting 2
          Gene Aliases:
          PMCA2, PMCA2a, PMCA2i
          Chromosome Location:
          Chr.3: 10324023 - 10707962 on Build GRCh38
          UniGene ID:
          Hs.268942
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 120230

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ATP2B2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001001331.2 NP_001001331.1 23 3918
          NM_001683.3 NP_001674.2 20 3783
          XM_005265179.4 XP_005265236.1 25 4102
          XM_006713175.3 XP_006713238.1 25 3986
          XM_011533752.2 XP_011532054.1 23 3891
          XM_017006481.1 XP_016861970.1 26 4229
          XM_017006482.1 XP_016861971.1 23 3920
          XM_017006483.1 XP_016861972.1 21 3930
          XM_017006484.1 XP_016861973.1 23 3893
          XM_017006485.1 XP_016861974.1 24 4197
          XM_017006486.1 XP_016861975.1 25 4252
          XM_017006487.1 XP_016861976.1 22 3850
          XM_017006488.1 XP_016861977.1 22 4104
          XM_017006489.1 XP_016861978.1 23 4026
          XM_017006490.1 XP_016861979.1 21 3944
          XM_017006491.1 XP_016861980.1 22 4116
          XM_017006492.1 XP_016861981.1 24 4111
          GenBank mRNA AB210008.1 - 20 3856
          DQ201777.1 - 12 1952
          DQ201778.1 - 8 1049
          L00620.1 - 19 3362
          L20977.1 - 22 3920
          M97260.1 - 20 3764
          S95758.1 - 19 3047
          X63575.1 - 22 3549

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          calcium ion transport
          organelle organization
          lactation
          sensory perception of sound
          locomotory behavior
          regulation of cell size
          cerebellar Purkinje cell differentiation
          cerebellar granule cell differentiation
          neuron differentiation
          ion transmembrane transport
          locomotion
          serotonin metabolic process
          otolith mineralization
          cGMP metabolic process
          regulation of synaptic plasticity
          synapse organization
          neuromuscular process controlling balance
          detection of mechanical stimulus involved in sensory perception of sound
          regulation of cytosolic calcium ion concentration
          positive regulation of calcium ion transport
          auditory receptor cell stereocilium organization
          calcium ion transmembrane transport
          cochlea development
          regulation of cardiac conduction
          calcium-transporting ATPase activity
          protein binding
          calmodulin binding
          ATP binding
          calcium-dependent ATPase activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0