Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RASGRP1 SIRNA Assays ›
          Gene Symbol:
          RASGRP1
          Gene Name:
          RAS guanyl releasing protein 1
          Gene Aliases:
          CALDAG-GEFI, CALDAG-GEFII, RASGRP, V, hRasGRP1
          Chromosome Location:
          Chr.15: 38488097 - 38564806 on Build GRCh38
          UniGene ID:
          Hs.591127
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 120435

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RASGRP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001128602.1 NP_001122074.1 2 256
          NM_001306086.1 NP_001293015.1 2 256
          NM_005739.3 NP_005730.2 2 256
          XM_005254114.2 XP_005254171.1 2 245
          XM_011521151.2 XP_011519453.1 2 245
          XM_017021860.1 XP_016877349.1 2 183
          GenBank mRNA AF081195.1 - 2 181
          AF081197.1 - 1 35
          AF106071.1 - 2 90
          AK295097.1 - 1 120
          AK314091.1 - 2 256
          AK316067.1 - 2 184
          AY634315.2 - 2 85
          AY858556.2 - 2 85
          AY954625.4 - 2 85
          AY966005.1 - 2 85
          BC067298.1 - 2 161
          BC109296.1 - 2 128 40034427
          BC109297.1 - 2 128 40034428
          DA993002.1 - 2 256

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          cytokine production
          positive regulation of protein phosphorylation
          inflammatory response to antigenic stimulus
          signal transduction
          Ras protein signal transduction
          regulation of phosphatidylinositol 3-kinase signaling
          cell differentiation
          secretory granule localization
          positive regulation of granulocyte macrophage colony-stimulating factor production
          positive regulation of tumor necrosis factor production
          positive regulation of natural killer cell differentiation
          positive regulation of T cell differentiation in thymus
          mast cell degranulation
          positive regulation of MAP kinase activity
          positive regulation of GTPase activity
          positive regulation of natural killer cell mediated cytotoxicity
          positive regulation of JNK cascade
          positive regulation of Ras protein signal transduction
          vesicle transport along microtubule
          protein oligomerization
          regulation of ERK1 and ERK2 cascade
          positive regulation of ERK1 and ERK2 cascade
          activation of GTPase activity
          regulation of phospholipase C activity
          regulation of p38MAPK cascade
          positive regulation of interferon-gamma secretion
          phosphatidylserine binding
          guanyl-nucleotide exchange factor activity
          Ras guanyl-nucleotide exchange factor activity
          calcium ion binding
          zinc ion binding
          lipid binding
          diacylglycerol binding
          phosphatidylcholine binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0