Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: HDAC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321225.1 | NP_001308154.1 | 5 | 440 | ||
NM_001321226.1 | NP_001308155.1 | 4 | 850 | |||
NM_001321227.1 | NP_001308156.1 | 4 | 571 | |||
NM_001321228.1 | NP_001308157.1 | 4 | 534 | |||
NM_001321229.1 | NP_001308158.1 | 4 | 388 | |||
NM_001321230.1 | NP_001308159.1 | 4 | 571 | |||
NM_001321231.1 | NP_001308160.1 | 4 | 850 | |||
NM_006044.3 | NP_006035.2 | 4 | 441 | |||
NR_135591.1 | 3 | 314 | ||||
NR_135592.1 | 3 | 448 | ||||
NR_135593.1 | 3 | 448 | ||||
XM_017029187.1 | XP_016884676.1 | 4 | 363 | |||
GenBank mRNA | AB020708.1 | - | 4 | 356 | ||
AF132609.1 | - | 3 | 284 | |||
AJ011972.1 | - | 4 | 377 | |||
AK024083.1 | - | 4 | 383 | |||
AK295884.1 | - | 3 | 301 | |||
AK302926.1 | - | 4 | 379 | |||
AK303012.1 | - | 2 | 282 | |||
AL137696.1 | - | 4 | 405 | |||
BC005872.1 | - | 4 | 810 | |||
BC011498.1 | - | 4 | 397 | |||
BC013737.1 | - | 3 | 266 | 2984860 | ||
BC069243.1 | - | 4 | 383 | 6584427 | ||
BT006649.1 | - | 3 | 284 | |||
DA191923.1 | - | 3 | 257 | |||
DC377684.1 | - | 4 | 441 | |||
DC402494.1 | - | 5 | 440 |